accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOSHI(E/Z)

CAS RN 179528-45-1

IUPAC Name: 3-(4-iodophenyl)-2-sulfanylprop-2-enoic acid
Synonyms: 3-(4-iodophenyl)-2-sulfanylprop-2-enoic acid
Molecular Weight (g/mol): 306.12
Molecular Formula: C9H7IO2S
InChi Key: DJCVSFWGKYHMKH-UHFFFAOYSA-N
SMILES: OC(=O)C(S)=CC1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)