CAS RN 1816254-69-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ DA ZP1
Fluorogenic Zn(II) sensor for isolation of stem cell derived β cells