accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1816290-25-1

NNNFFFFF

CAS RN 1816290-25-1

IUPAC Name: 5-(1,1,2,2,2-pentafluoroethyl)pyrimidine-2-carbonitrile
Synonyms: 5-(1,1,2,2,2-pentafluoroethyl)pyrimidine-2-carbonitrile
Molecular Weight (g/mol): 223.11
Molecular Formula: C7H2F5N3
InChi Key: GGNFRHPEYLAGFK-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C1=CN=C(N=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results