CAS RN 181695-72-7
IUPAC Name:
4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzene-1-sulfonamide
Synonyms:
valdecoxib COX valdyn bextra valecoxib kudeq
Molecular Weight (g/mol):
314.36
Molecular Formula:
C16H14N2O3S
InChi Key:
LNPDTQAFDNKSHK-UHFFFAOYSA-N
SMILES:
CC1=C(C(=NO1)C1=CC=CC=C1)C1=CC=C(C=C1)S(N)(=O)=O
1
–
2
of
2
results
1
R&D Systems™ Valdecoxib
CAS: 181695-72-7 Molecular Formula: C16H14N2O3S
2
Thermo Scientific Chemicals Valdecoxib
CAS: 181695-72-7 Molecular Formula: C16H14N2O3S Molecular Weight (g/mol): 314.46