CAS RN 188591-46-0
IUPAC Name:
4-chloro-N-(2-{[5-(trifluoromethyl)pyridin-2-yl]sulfonyl}ethyl)benzamide
Synonyms:
4-chloro-N-{2-[5-(trifluoromethyl)pyridin-2-ylsulfonyl]ethyl}benzamide
Molecular Weight (g/mol):
392.78
Molecular Formula:
C15H12ClF3N2O3S
InChi Key:
JFUIMTGOQCQTPF-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CN=C(C=C1)S(=O)(=O)CCNC(=O)C1=CC=C(Cl)C=C1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ PPARβ/δ Antagonist, GSK3787
The PPARβ/δAntagonist II, PT-S58, also referenced under CAS 188591-46-0, controls the biological activity of PPARβ/δ. This small molecule/inhibitor is primarily used for Biochemicals applications.
2
R&D Systems™ GSK 3787
CAS: 188591-46-0 Molecular Formula: C15H12ClF3N2O3S