accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OBrNHClN

CAS RN 191034-25-0

IUPAC Name: 4-[3-(5-bromo-2-propoxyphenyl)-5-(4-chlorophenyl)-1H-pyrrol-2-yl]pyridine
Synonyms: 4-[3-(5-bromo-2-propoxyphenyl)-5-(4-chlorophenyl)-1H-pyrrol-2-yl]pyridine
Molecular Weight (g/mol): 467.79
Molecular Formula: C24H20BrClN2O
InChi Key: HHBOWXZOLYQFNY-UHFFFAOYSA-N
SMILES: CCCOC1=C(C=C(Br)C=C1)C1=C(NC(=C1)C1=CC=C(Cl)C=C1)C1=CC=NC=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results