accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3H3COOHNOHOHOOOOOOH3COOH3CNHOOOHOOOHCH3OOH3C(?)

CAS RN 191930-58-2

IUPAC Name: 5-[(1-{[4,12-bis(acetyloxy)-2-(benzoyloxy)-1-hydroxy-15-{[2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-9-yl]oxy}-1-oxopropan-2-yl)carbamoyl]-2-(6-hydroxy-3-oxo-3H-xanthen-9-yl)benzoic acid
Synonyms: 5-[(1-{[4,12-bis(acetyloxy)-2-(benzoyloxy)-1-hydroxy-15-{[2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-9-yl]oxy}-1-oxopropan-2-yl)carbamoyl]-2-(3-hydroxy-6-oxoxanthen-9-yl)benzoic acid
Molecular Weight (g/mol): 1283.3
Molecular Formula: C71H66N2O21
InChi Key: JMWJJMCMRRQCSY-UHFFFAOYNA-N
SMILES: CC(NC(=O)C1=CC(C(O)=O)=C(C=C1)C1=C2C=CC(=O)C=C2OC2=CC(O)=CC=C12)C(=O)OC1CC2OCC2(OC(C)=O)C2C(OC(=O)C3=CC=CC=C3)C3(O)CC(OC(=O)C(O)C(NC(=O)C4=CC=CC=C4)C4=CC=CC=C4)C(C)=C(C(OC(C)=O)C(=O)C12C)C3(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)