CAS RN 192441-08-0
IUPAC Name:
6-[(4-bromothiophen-2-yl)methoxy]-7H-purin-2-amine
Synonyms:
lomeguatrib
Molecular Weight (g/mol):
326.17
Molecular Formula:
C10H8BrN5OS
InChi Key:
JUJPKFNFCWJBCX-UHFFFAOYSA-N
SMILES:
NC1=NC(OCC2=CC(Br)=CS2)=C2NC=NC2=N1