accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHHOO(E)

CAS RN 19309-14-9

IUPAC Name: (2E)-1-(2,4-dihydroxy-6-methoxyphenyl)-3-phenylprop-2-en-1-one
Synonyms: cardamonin
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: NYSZJNUIVUBQMM-BQYQJAHWSA-N
SMILES: COC1=CC(O)=CC(O)=C1C(=O)\C=C\C1=CC=CC=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results