Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cardamonin
SDP

Catalog No. 250950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-095-0 50 mg
25-091-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-095-0 Supplier Tocris Bioscience™ Supplier No. 2509/50

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of NF-κB activation; anti-inflammatory

Chalone analog that display anti-inflammatory activity. Inhibits NO and PGE2 production from LPS- and IFN-γ-induced RAW cells and inhibits TXB2 production via the COX-1 and COX-2 pathways. Inhibits NF-κB activation via inhibition of IκBα degradation and phosphorylation, IκB kinase activation and NF-κB nuclear translocation.

Chemical Identifiers

CAS 19309-14-9
Molecular Formula C16H14O4

Specifications

Quantity 50 mg
Formula Weight 270.28
Purity >98%
Molecular Weight (g/mol) 270.284
Chemical Name or Material Cardamonin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.