CAS RN 1992047-63-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ M 084 hydrochloride
TRPC4/5 channel blocker; antidepressant and anxiolytic