accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19983-44-9

IUPAC Name: 6-nitro-1λ⁶-benzothiophene-1,1-dione
Synonyms: 6-nitro-1λ⁶-benzothiophene-1,1-dione
Molecular Weight (g/mol): 211.19
Molecular Formula: C8H5NO4S
InChi Key: ZRRGOUHITGRLBA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=C(C=CS2(=O)=O)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results