CAS RN 200006-08-2
IUPAC Name:
2-[(3-{[1,1'-biphenyl]-4-yloxy}-3-(4-fluorophenyl)propyl)(methyl)amino]acetic acid hydrochloride
Synonyms:
[(3-{[1,1'-biphenyl]-4-yloxy}-3-(4-fluorophenyl)propyl)(methyl)amino]acetic acid hydrochloride
Molecular Weight (g/mol):
429.92
Molecular Formula:
C24H25ClFNO3
InChi Key:
RPDGSZCYSJWQEE-UHFFFAOYNA-N
SMILES:
Cl.CN(CCC(OC1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(F)C=C1)CC(O)=O
1
–
1
of
1
results
1
Tocris Bioscience™ ALX 5407 hydrochloride
CAS: 200006-08-2 Molecular Formula: C24H25ClFNO3