accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 20153-98-4

HHClClH3COOH3COH3COONNOOOCH3OCH3OH3C

CAS RN 20153-98-4

IUPAC Name: dihydrogen 3-{4-[3-(3,4,5-trimethoxybenzoyloxy)propyl]-1,4-diazepan-1-yl}propyl 3,4,5-trimethoxybenzoate dichloride
Synonyms: dihydrogen dilazep dichloride
Molecular Weight (g/mol): 677.61
Molecular Formula: C31H46Cl2N2O10
InChi Key: VILIWRRWAWKXRW-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC(OC)=C1OC)C(=O)OCCCN1CCCN(CCCOC(=O)C2=CC(OC)=C(OC)C(OC)=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results