CAS RN 2016864-46-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ AZ 1729
FFA2 allosteric agonist; displays bias for the Gi signaling pathway