CAS RN 204326-43-2
IUPAC Name:
12-bromo-4,8-dioxa-2,5-diazatricyclo[7.4.0.0²,⁶]trideca-1(9),5,10,12-tetraen-3-one
Synonyms:
12-bromo-4,8-dioxa-2,5-diazatricyclo[7.4.0.0²,⁶]trideca-1(9),5,10,12-tetraen-3-one
Molecular Weight (g/mol):
269.05
Molecular Formula:
C9H5BrN2O3
InChi Key:
MUDRLQRJCGJJTB-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(OCC3=NOC(=O)N23)C=C1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ NS 2028
Potent, specific, and irreversible inhibitor of soluble guanylyl cyclase.
2
R&D Systems™ NS 2028
CAS: 204326-43-2 Molecular Formula: C9H5BrN2O3