CAS RN 2253744-54-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Dooku 1
Reversibly blocks Yoda1 activity; does not affect constitutive Piezo1 activity