CAS RN 2262452-06-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ RU.521
Potent and selective inhibitor of cyclic GMP-AMP synthase (cGAS)