accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOOHNONHOH3CCH3H3CCH3

CAS RN 230961-21-4

IUPAC Name: 3-({1-[(2-methoxy-1-phenylethyl)carbamoyl]-2,2-dimethylpropyl}carbamoyl)-6-{2-methyl-[1,1'-biphenyl]-4-yl}hexanoic acid
Synonyms: 3-({1-[(2-methoxy-1-phenylethyl)carbamoyl]-2,2-dimethylpropyl}carbamoyl)-6-{2-methyl-[1,1'-biphenyl]-4-yl}hexanoic acid
Molecular Weight (g/mol): 572.75
Molecular Formula: C35H44N2O5
InChi Key: NSMABJUGSNPHMN-UHFFFAOYNA-N
SMILES: COCC(NC(=O)C(NC(=O)C(CCCC1=CC(C)=C(C=C1)C1=CC=CC=C1)CC(O)=O)C(C)(C)C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)