accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 232605-26-4

CH2OO

CAS RN 232605-26-4

IUPAC Name: 5-methylidene-6a-[(naphthalen-2-yl)methyl]-hexahydro-1H-cyclopenta[c]furan-1-one
Synonyms: 5-methylidene-6a-(naphthalen-2-ylmethyl)-tetrahydrocyclopenta[c]furan-1-one
Molecular Weight (g/mol): 278.35
Molecular Formula: C19H18O2
InChi Key: CVIRWLJKDBYYOG-UHFFFAOYNA-N
SMILES: C=C1CC2COC(=O)C2(CC2=CC=C3C=CC=CC3=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)