Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Bay 36-7620
GREENER_CHOICESDP

Catalog No. 250110 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-011-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-011-0 Supplier R&D Systems™ Supplier No. 2501/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. mGlu1 inverse agonist

Selective mGlu1 receptor non-competitive antagonist (IC50 = 0.16 μM) with inverse agonist activity. Impairs classical conditioning and associated synaptic plasticity in hippocampal neurons. Exhibits neuroprotective and anticonvulsive effects in vivo following systemic administration.

Chemical Identifiers

CAS 232605-26-4
Molecular Formula C19H18O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 278.35
Purity >98%
Molecular Weight (g/mol) 278.351
Chemical Name or Material Bay 36-7620
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.