accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHH2OClClH2NOHH2NHOO

CAS RN 23279-22-3

IUPAC Name: dihydrogen 2-amino-3-(3-amino-4-hydroxyphenyl)propanoic acid hydrate dichloride
Synonyms: dihydrogen 2-amino-3-(3-amino-4-hydroxyphenyl)propanoic acid hydrate dichloride
Molecular Weight (g/mol): 287.14
Molecular Formula: C9H16Cl2N2O4
InChi Key: ITQBVYBSXFBTJX-UHFFFAOYNA-N
SMILES: [H+].[H+].O.[Cl-].[Cl-].NC(CC1=CC=C(O)C(N)=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)