accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23563-26-0

H3CONHCH3O

CAS RN 23563-26-0

IUPAC Name: N-(4-butoxyphenyl)acetamide
Synonyms: N-(4-butoxyphenyl)acetamide
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: JYMFGHFNWSMPBP-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(NC(C)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results