Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butoxyacetanilide, 97%, Thermo Scientific™

Catalog No. AC326260050
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23563-26-0
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.27
MDL Number MFCD00027197
InChI Key JYMFGHFNWSMPBP-UHFFFAOYSA-N
Synonym n-4-butoxyphenyl acetamide, acetamide, n-4-butoxyphenyl, 4'-n-butoxyacetanilide, o-butyl paracetamol
PubChem CID 90165
IUPAC Name N-(4-butoxyphenyl)acetamide
SMILES CCCCOC1=CC=C(C=C1)NC(=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 110°C to 112°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula CH3(CH2)3OC6H4NHCOCH3
Quantity 5 g
Formula Weight 207.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Butoxyacetanilide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 245-739-1
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.