accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 235788-61-1

OHOOOOHONNHH3CH3CCH3(E)

CAS RN 235788-61-1

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-[(E)-[1-(2-hydroxy-4,4-dimethyl-6-oxocyclohex-1-en-1-yl)ethylidene]amino]butanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-[(E)-[1-(2-hydroxy-4,4-dimethyl-6-oxocyclohex-1-en-1-yl)ethylidene]amino]butanoic acid
Molecular Weight (g/mol): 504.58
Molecular Formula: C29H32N2O6
InChi Key: AFBHGOUICAXOQZ-OCSSWDANSA-N
SMILES: C\C(=N/CCC(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)C1=C(O)CC(C)(C)CC1=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)