CAS RN 2362575-45-7
CAS RN 2362575-45-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ CRBN-6-5-5-VHL
Potent and selective cereblon Degrader (PROTAC™); cell-permeable