CAS RN 2455450-63-0
IUPAC Name:
6-(5-{[(2S)-azetidin-2-yl]methoxy}pyridin-3-yl)hex-5-yn-1-ol dihydrochloride
Synonyms:
sazetidine A
Molecular Weight (g/mol):
333.25
Molecular Formula:
C15H22Cl2N2O2
InChi Key:
WRSKKJWUXISFFU-UTLKBRERSA-N
SMILES:
Cl.Cl.OCCCCC#CC1=CN=CC(OC[C@@H]2CCN2)=C1
1
–
1
of
1
results
1
R&D Systems™ Sazetidine A dihydrochloride
CAS: 2455450-63-0 Molecular Formula: C15H22Cl2N2O2