accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2NNNNF

CAS RN 256376-24-6

IUPAC Name: 5-cyclopropyl-2-{1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl}pyrimidin-4-amine
Synonyms: 5-cyclopropyl-2-{1-[(2-fluorophenyl)methyl]pyrazolo[3,4-b]pyridin-3-yl}pyrimidin-4-amine
Molecular Weight (g/mol): 360.4
Molecular Formula: C20H17FN6
InChi Key: ATOAHNRJAXSBOR-UHFFFAOYSA-N
SMILES: NC1=NC(=NC=C1C1CC1)C1=NN(CC2=CC=CC=C2F)C2=NC=CC=C12
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results