Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BAY 41-2272
GREENER_CHOICESDP

Catalog No. 443050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-305-0 50 mg
44-301-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-305-0 Supplier R&D Systems™ Supplier No. 4430/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Soluble guanylyl cyclase (sGC) activator

Activator of soluble guanylyl cyclase (sGC); acts at a nitric oxide (NO)-independent regulatory site in the sGC α1 subunit. Inhibits platelet aggregation (IC50 = 36 nM) and phenylephrine-induced contractions of rabbit aorta (IC50 = 0.30 μM). Also reduces vascular smooth muscle growth through cAMP- and cGMP-dependent PKA and PKG pathways.

Chemical Identifiers

CAS 256376-24-6
Molecular Formula C20H17FN6

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 360.39
Purity >98%
Molecular Weight (g/mol) 360.40
Chemical Name or Material BAY 41-2272
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.