accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNH2NNNNFNH2

CAS RN 256498-66-5

IUPAC Name: 2-{1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl}-5-(morpholin-4-yl)pyrimidine-4,6-diamine
Synonyms: 2-{1-[(2-fluorophenyl)methyl]pyrazolo[3,4-b]pyridin-3-yl}-5-(morpholin-4-yl)pyrimidine-4,6-diamine
Molecular Weight (g/mol): 420.45
Molecular Formula: C21H21FN8O
InChi Key: AQYFUZRYBJBAGZ-UHFFFAOYSA-N
SMILES: NC1=NC(=NC(N)=C1N1CCOCC1)C1=NN(CC2=CC=CC=C2F)C2=NC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results