accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOHNCH3H3CCH3H3CNOHO

CAS RN 260264-93-5

IUPAC Name: N-[2,2-dimethyl-1-(methylcarbamoyl)propyl]-2-[1-(N-hydroxyformamido)ethyl]-5-phenylpentanamide
Synonyms: N-[2,2-dimethyl-1-(methylcarbamoyl)propyl]-2-[1-(N-hydroxyformamido)ethyl]-5-phenylpentanamide
Molecular Weight (g/mol): 391.51
Molecular Formula: C21H33N3O4
InChi Key: GHVMTHKJUAOZJP-UHFFFAOYNA-N
SMILES: CNC(=O)C(NC(=O)C(CCCC1=CC=CC=C1)C(C)N(O)C=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results