CAS RN 264618-38-4
IUPAC Name:
1-(1-{3-[1-benzoyl-3-(3,4-dichlorophenyl)piperidin-3-yl]propyl}-4-phenylpiperidin-4-yl)-3,3-dimethylurea hydrochloride
Synonyms:
1-(1-{3-[1-benzoyl-3-(3,4-dichlorophenyl)piperidin-3-yl]propyl}-4-phenylpiperidin-4-yl)-3,3-dimethylurea hydrochloride
Molecular Weight (g/mol):
658.11
Molecular Formula:
C35H43Cl3N4O2
InChi Key:
HYPKKQPFHSNZBY-UHFFFAOYNA-N
SMILES:
Cl.CN(C)C(=O)NC1(CCN(CCCC2(CCCN(C2)C(=O)C2=CC=CC=C2)C2=CC(Cl)=C(Cl)C=C2)CC1)C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ SSR 146977 hydrochloride
CAS: 264618-38-4 Molecular Formula: C35H43Cl3N4O2