accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONOCH3HNNOONHN

CAS RN 264622-58-4

IUPAC Name: N-(4-cyanophenyl)-2-[4-(2,6-dioxo-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purin-8-yl)phenoxy]acetamide
Synonyms: N-(4-cyanophenyl)-2-[4-(2,6-dioxo-1,3-dipropyl-7H-purin-8-yl)phenoxy]acetamide
Molecular Weight (g/mol): 486.53
Molecular Formula: C26H26N6O4
InChi Key: AJBBEYXFRYFVNM-UHFFFAOYSA-N
SMILES: CCCN1C2=C(NC(=N2)C2=CC=C(OCC(=O)NC3=CC=C(C=C3)C#N)C=C2)C(=O)N(CCC)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)