CAS RN 293-96-9
IUPAC Name:
cyclodecane
Synonyms:
cyclodecane
Molecular Weight (g/mol):
140.27
Molecular Formula:
C10H20
InChi Key:
LMGZGXSXHCMSAA-UHFFFAOYSA-N
SMILES:
C1CCCCCCCCC1
1
–
2
of
2
results
1
MP Biomedicals, Inc Cyclodecane, MP Biomedicals
CAS: 293-96-9 Molecular Formula: C10H20 Molecular Weight (g/mol): 140.27 InChI Key: LMGZGXSXHCMSAA-UHFFFAOYSA-N Synonym: cyclodecan,cyclodecyl,acmc-1cklb,cyclodecane PubChem CID: 9267 IUPAC Name: cyclodecane SMILES: C1CCCCCCCCC1
2
Cyclodecane 94.0+%, TCI America™
CAS: 293-96-9 Molecular Formula: C10H20 Molecular Weight (g/mol): 140.27 MDL Number: MFCD00003711 InChI Key: LMGZGXSXHCMSAA-UHFFFAOYSA-N Synonym: cyclodecan,cyclodecyl,acmc-1cklb,cyclodecane PubChem CID: 9267 IUPAC Name: cyclodecane SMILES: C1CCCCCCCCC1