accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNSCH3HBr

CAS RN 298197-04-3

IUPAC Name: 4-[4-(4-methylphenyl)-1,3-thiazol-2-yl]-2-propylpyridine hydrobromide
Synonyms: 4-[4-(4-methylphenyl)-1,3-thiazol-2-yl]-2-propylpyridine hydrobromide
Molecular Weight (g/mol): 375.33
Molecular Formula: C18H19BrN2S
InChi Key: RJCFNQZVFUMORB-UHFFFAOYSA-N
SMILES: Br.CCCC1=NC=CC(=C1)C1=NC(=CS1)C1=CC=C(C)C=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results