CAS RN 300803-69-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ OB-1
Stomatin-like protein-3 (STOML3) oligomerization inhibitor