accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNSOH2NOCH3

CAS RN 301305-73-7

IUPAC Name: 2-(3,4-dimethoxybenzamido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Synonyms: 2-(3,4-dimethoxybenzamido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Molecular Weight (g/mol): 360.43
Molecular Formula: C18H20N2O4S
InChi Key: FSPQCTGGIANIJZ-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C=C1)C(=O)NC1=C(C(N)=O)C2=C(CCCC2)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Product Type 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results