accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30429-72-2

CH3CH3HOCH3CH3HOCH3CH3HOHo

CAS RN 30429-72-2

IUPAC Name: tris(propan-2-ol) holmium
Synonyms: tris(isopropyl alcohol) holmium
Molecular Weight (g/mol): 345.22
Molecular Formula: C9H24HoO3
InChi Key: XTBDKTFYOKTZRX-UHFFFAOYSA-N
SMILES: [Ho].CC(C)O.CC(C)O.CC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results