Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Holmium(III) isopropoxide, 5% w/v in toluene/isopropanol, Thermo Scientific™
Click to view available options
Quantity:
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | Red |
| Linear Formula | Ho[OCH(CH3)2]3 |
| Quantity | 100 mL |
| UN Number | UN1993 |
| Sensitivity | Moisture Sensitive |
| Formula Weight | 342.19 |
| Concentration or Composition (by Analyte or Components) | 5% w/v in toluene/isopropanol |
| Physical Form | Liquid |
| Chemical Name or Material | Holmium(III) isopropoxide |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H225-H304-H361-H373-H314-H336 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. |
| Precautionary Statement | GHS P Statement P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a Keep away from heat/sparks/open flames/hot surfaces. No smoking. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction