Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Spermine tetrahydrochloride
GREENER_CHOICESDP

Catalog No. 0958100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-581-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-581-00 Supplier R&D Systems™ Supplier No. 0958/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Modulator at polyamine site

Polyamine which produces a variety of modulatory effects on the NMDA receptor channel, acting through a specific site on the complex which can cause both agonist and antagonist effects.

Chemical Identifiers

CAS 306-67-2
Molecular Formula C10H30Cl4N4

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 348.19
Purity >98%
Molecular Weight (g/mol) 348.18
Chemical Name or Material Spermine tetrahydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.