accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30754-23-5

OOOHOHOOOHOHOOOHOOHOOHOOHOOOHOHOOOOHOHIIIIIIHOOHI

CAS RN 30754-23-5

IUPAC Name: 5,10,15,20,25,30,35-heptakis(iodomethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
Synonyms: 5,10,15,20,25,30,35-heptakis(iodomethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
Molecular Weight (g/mol): 1904.27
Molecular Formula: C42H63I7O28
InChi Key: YODKQJPYQJRHEQ-UHFFFAOYNA-N
SMILES: OC1C(O)C2OC3OC(CI)C(OC4OC(CI)C(OC5OC(CI)C(OC6OC(CI)C(OC7OC(CI)C(OC8OC(CI)C(OC1OC2CI)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results