CAS RN 312917-14-9
IUPAC Name:
N-(3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl)naphthalene-1-carboxamide
Synonyms:
N-(3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl)naphthalene-1-carboxamide
Molecular Weight (g/mol):
332.42
Molecular Formula:
C20H16N2OS
InChi Key:
WQGDQGAFSDMBLA-UHFFFAOYSA-N
SMILES:
O=C(NC1=C(C#N)C2=C(CCCC2)S1)C1=C2C=CC=CC2=CC=C1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ JNK Inhibitor IX
The JNK Inhibitor IX, also referenced under CAS 312917-14-9, controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
2
R&D Systems™ TCS JNK 5a
CAS: 312917-14-9 Molecular Formula: C20H16N2OS