accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHNSClClOOHNOO(E/Z)

CAS RN 315706-13-9

IUPAC Name: 2-{2-[4-(3,4-dichlorophenyl)-1,3-thiazol-2-yl]hydrazin-1-ylidene}-3-(2-nitrophenyl)propanoic acid
Synonyms: 2-{2-[4-(3,4-dichlorophenyl)-1,3-thiazol-2-yl]hydrazin-1-ylidene}-3-(2-nitrophenyl)propanoic acid
Molecular Weight (g/mol): 451.28
Molecular Formula: C18H12Cl2N4O4S
InChi Key: KFRKRECSIYXARE-UHFFFAOYSA-N
SMILES: OC(=O)C(CC1=CC=CC=C1[N+]([O-])=O)=NNC1=NC(=CS1)C1=CC(Cl)=C(Cl)C=C1
Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)