accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNBrH2NCl

CAS RN 321351-00-2

IUPAC Name: 4-bromo-N1-[3-chloro-4-(2-methylbenzoyl)phenyl]benzene-1,2-diamine
Synonyms: 4-bromo-N1-[3-chloro-4-(2-methylbenzoyl)phenyl]benzene-1,2-diamine
Molecular Weight (g/mol): 415.72
Molecular Formula: C20H16BrClN2O
InChi Key: HDCLCHNAEZNGNV-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(=O)C1=C(Cl)C=C(NC2=C(N)C=C(Br)C=C2)C=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results