accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32988-50-4

IUPAC Name: (3S)-3,6-diamino-N-[(3S,6Z,9S,12S,15S)-3-[(4R,6S)-2-amino-6-hydroxy-3,4,5,6-tetrahydropyrimidin-4-yl]-6-[(carbamoylamino)methylidene]-9,12-bis(hydroxymethyl)-2,5,8,11,14-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-15-yl]hexanamide
Synonyms: viomycin
Molecular Weight (g/mol): 685.7
Molecular Formula: C25H43N13O10
InChi Key: GXFAIFRPOKBQRV-GHXCTMGLSA-N
SMILES: NCCC[C@H](N)CC(=O)N[C@H]1CNC(=O)[C@@H](NC(=O)\C(NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC1=O)=C\NC(N)=O)[C@H]1C[C@H](O)N=C(N)N1
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results