accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHNSNHNNH2N

CAS RN 333962-91-7

IUPAC Name: 2-amino-6-{[(1H-imidazol-2-yl)methyl]sulfanyl}-4-(4-oxocyclohexa-2,5-dien-1-ylidene)-1,4-dihydropyridine-3,5-dicarbonitrile
Synonyms: 2-amino-6-[(1H-imidazol-2-ylmethyl)sulfanyl]-4-(4-oxocyclohexa-2,5-dien-1-ylidene)-1H-pyridine-3,5-dicarbonitrile
Molecular Weight (g/mol): 348.38
Molecular Formula: C17H12N6OS
InChi Key: LOLJUQOCQBYBGT-UHFFFAOYSA-N
SMILES: NC1=C(C#N)C(=C2C=CC(=O)C=C2)C(C#N)=C(N1)SCC1=NC=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results