CAS RN 338736-46-2
IUPAC Name:
6-methoxy-N-(4-methoxyphenyl)quinazolin-4-amine hydrochloride
Synonyms:
6-methoxy-N-(4-methoxyphenyl)quinazolin-4-amine hydrochloride
Molecular Weight (g/mol):
317.77
Molecular Formula:
C16H16ClN3O2
InChi Key:
AVKFOWUSTVWZQN-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC=C(NC2=C3C=C(OC)C=CC3=NC=N2)C=C1
1
–
1
of
1
results
1
R&D Systems™ LY 456236 hydrochloride
CAS: 338736-46-2 Molecular Formula: C16H16ClN3O2