Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Sunitinib malate
GREENER_CHOICESDP

Catalog No. 376850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-685-0 50 mg
37-681-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-685-0 Supplier R&D Systems™ Supplier No. 3768/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent VEGFR, PDGFRβ and KIT inhibitor

Potent, ATP-competitive VEGFR, PDGFRβ and KIT inhibitor (Ki values are 2, 9, 17, 8 and 4 nM for VEGFR -1, -2, -3, PDGFRβ and KIT respectively). Also inhibits cellular receptor phosphorylation of FLT3, RET and CSF-1R. Exhibits antiangiogenic and antitumor activity in multiple xenograft models.

Chemical Identifiers

CAS 341031-54-7
Molecular Formula C26H33FN4O7

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 532.56
Purity >99%
Molecular Weight (g/mol) 532.569
Chemical Name or Material Sunitinib malate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.