CAS RN 343630-41-1
CAS RN 343630-41-1
IUPAC Name:
1-[4-bromo-2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]-3-[3-(trifluoromethyl)phenyl]urea
Synonyms:
1-[4-bromo-2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]-3-[3-(trifluoromethyl)phenyl]urea
Molecular Weight (g/mol):
427.19
Molecular Formula:
C15H10BrF3N6O
InChi Key:
JXPULDIATMTIIN-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(NC(=O)NC2=C(C=C(Br)C=C2)C2=NNN=N2)=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ NS 3623


CAS: 343630-41-1 Molecular Formula: C15H10BrF3N6O Molecular Weight (g/mol): 427.185