accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONNNNNH2

CAS RN 343787-29-1

IUPAC Name: 1-{2-[5-(2-methoxyethoxy)-1H-1,3-benzodiazol-1-yl]quinolin-8-yl}piperidin-4-amine
Synonyms: 1-{2-[5-(2-methoxyethoxy)-1,3-benzodiazol-1-yl]quinolin-8-yl}piperidin-4-amine
Molecular Weight (g/mol): 417.51
Molecular Formula: C24H27N5O2
InChi Key: DEEOXSOLTLIWMG-UHFFFAOYSA-N
SMILES: COCCOC1=CC=C2N(C=NC2=C1)C1=CC=C2C=CC=C(N3CCC(N)CC3)C2=N1
Product Type 
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)